missing translation for 'onlineSavingsMsg'
Learn More

Buflomedil hydrochloride, MedChemExpress

Product Code. 16448336
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16448336

Brand: Medchem Express HYB0484

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Buflomedil hydrochloride is a vasodilator used to treat claudication or the symptoms of peripheral arterial disease.

Buflomedil hydrochloride is a vasodilator used to treat claudication or the symptoms of peripheral arterial disease.

Specifications

Chemical Name or Material Buflomedil hydrochloride
CAS 35543-24-9
Color White
Purity Grade Notes Research
Molecular Formula C17H26ClNO4
Quantity 100 mg
Solubility Information H2O : ≥ 55 mg/mL (159.95 mM) ∣DMSO : 33.33 mg/mL (96.93 mM; Need ultrasonic)
SMILES O=C(C1=C(OC)C=C(OC)C=C1OC)CCCN2CCCC2.Cl
Molecular Weight (g/mol) 343.85
Formula Weight 343.85
Percent Purity 99.67%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.