missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Buffers for NucleoSpin™ Virus Kits

Product Code. 15849137
Change view
Click to view available options
Quantity:
120 mL
200 mL
40 mL RAV1 + 1.0 mg Carrier RNA
Chemical Name or Material:
Lysis Buffer RAV1
Lysis Buffer RAV1 + Carrier RNA
Wash Buffer RAV3
Unit Size:
120mL
200mL
40mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Chemical Name or Material unitSize
15849137 120 mL Lysis Buffer RAV1 120mL
15869137 40 mL RAV1 + 1.0 mg Carrier RNA Lysis Buffer RAV1 + Carrier RNA 40mL
15859137 200 mL Wash Buffer RAV3 200mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15849137 Supplier Macherey-Nagel™ Supplier No. 740925
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Supplemental buffer solutions (e.g., wash and lysis buffers) for use with NucleoSpin™ Virus kits for the rapid isolation of viral nucleic acids (RNA and DNA).

Compatible kits include:

  • NucleoSpin™ RNA Virus
  • NucleoSpin™ RNA Virus F
  • NucleoSpin™ Dx Virus
  • NucleoSpin™ 8/96 Virus

Specifications

Chemical Name or Material Lysis Buffer RAV1
CAS 593-84-0
Color Colorless
For Use With (Application) Isolation of viral RNA and DNA
For Use With (Equipment) Compatible NucleoSpin Virus kits
pH 7.0 to 7.5
Physical Form Liquid
Quantity 120 mL
Formulation Guanidinium thiocyanate; Water
Odor Odorless
compliance-icons
Product Identifier
  • Buffer RAV1 (120 mL)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Supplemental information
  • MIXTURE LIST-Contain: guanidine thiochloride, triton x-100
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.