missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Buffers for NucleoSpin™ Plasmid Kits
Description
For use with the following NucleoSpin™ Plasmid DNA purification kits:
- NucleoSpin™ Plasmid (Lid/NoLid)
- NucleoSpin™ Plasmid QuickPure
- NucleoSpin™ Plasmid EasyPure
- NucleoSpin™ 8/96 Plasmid (Full/Core Kit)
- NucleoSpin™ Plasmid Transfection-Grade
- NucleoSpin™ 96 Plasmid Transfection-Grade (Full/Core Kit)
Specifications
Specifications
| Chemical Name or Material | Elution Buffer AE |
| Color | Colorless |
| For Use With (Application) | Preparation high-purity plasmid DNA |
| For Use With (Equipment) | NucleoSpin™ Plasmid kits |
| pH | 8.0 to 9.0 |
| Physical Form | Liquid |
| Quantity | 3000 mL |
| Odor | Odorless |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?