missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Buffers for NucleoSpin™ 96 DNA Plasma

Product Code. 15809127
Click to view available options
Quantity:
125 mL
250 mL
Chemical Name or Material:
Binding buffer PMB
Lysis buffer PML
Unit Size:
125mL
250mL
This item is not returnable. View return policy

Product Code. 15809127

Brand: Macherey-Nagel™ 740835.125

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Additional supply of buffer solutions for use in isolation of cell-free, circulating DNA from up to 2 mL blood plasma in high-throughput 96-well format using the NucleoSpin™ 96 DNA kit or core kit.

NucleoSpin™ 96 DNA Plasma

  • Isolation of cell-free DNA from up to 2 mL blood plasma (i.e., Liquid Biopsies)
  • Highly efficient recovery of cell-free DNA
  • High throughput 96-well format solution
  • Manual or automated processing
  • Optimized protocol for Cell-Free DNA BCT (Streck Tubes)
  • Core Kits contain basic content focused on automation platforms
  • Typical downstream applications: NGS, qPCR, ddPCR

Specifications

Chemical Name or Material Lysis buffer PML
CAS 50-01-1
Color Colorless
For Use With (Application) Isolation of cell-free DNA from plasma 
For Use With (Equipment) NucleoSpin™ 96 DNA Plasma kit or core kit
pH 4.5 to 5.2
Physical Form Liquid
Quantity 125 mL
Formulation Guanidine hydrochloride, Water
Odor Odorless
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.