missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Buffered Peptone Water ISO Formulation, Dehydrated

Product Code. 17933062
Change view
Click to view available options
Weight (Metric):
500 g
5 kg
Unit Size:
5kg
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (Metric) unitSize
17933062 500 g Each
17269969 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17933062 Supplier Thermo Scientific™ Supplier No. CM1211B

Please to purchase this item. Need a web account? Register with us today!

Pre-enrich food samples prior to the isolation of Salmonella species using this ISO compliant pre-enrichment medium.

Pre-enrich food samples prior to the isolation of Salmonella species using Thermo Scientific™ Oxoid™ Buffered Peptone Water ISO Formulation (Dehydrated). Buffered Peptone Water is a non-selective pre-enrichment medium and is tested in accordance with ISO 65791 and ISO 11133:20142. It also provides conditions for resuscitation of cells that have been injured by processes of food preservation

Low numbers and/or sub-lethally injured Salmonella cells may be present in a test food sample. Pre-enrichment in non-selective Buffered Peptone Water allows the cells time to repair and multiply before being introduced into selective culture, improving the chances of recovering Salmonella species from the sample.

  • Supports high growth and recovery rate: rich in nutrients, the medium produces high resuscitation rates and supports intense growth. Differs from Buffered Peptone water (ISO) CM1049 by the inclusion of a beef peptone.
  • Helps recovery of sub-lethally damaged salmonellae: the phosphate buffer system of the medium prevents bacterial damage due to changes in pH of the medium and helps the recovery of sub-lethally damaged salmonellae that may be sensitive to low pH.
  • ISO compliant: tested in accordance with ISO 65791and ISO 11133:20142.
TRUSTED_SUSTAINABILITY

Specifications

Description Buffered Peptone Water ISO Formulation (Dehydrated)
Form Powder
Product Type Pre-enrichment Medium
Quantity Each
Weight (Metric) 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.