missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- You can eliminate the time and expense of preparation and standardisation of solutions, helping streamline high-volume analytical work in applications such as quality control
- Contain 0.01% Sodium benzoate
- Certifications: This buffer is traceable to NIST Standard reference buffer
Specifications
Specifications
| Certificate of Analysis | Traceable to NIST Standard reference buffer |
| pH | 7.00 |
| Quantity | 1 L |
| Packaging | Plastic bottle |
| Chemical Name or Material | Phosphate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?