missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH8.00 (Borate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™

Brand:  Fisher Chemical J/2860/17

134.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.



Code : 23

Additional Details : Weight : 2.50000kg


 View more versions of this product

Product Code. 10141620

  • £171.00 / 2.50 L
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Buffer Solution pH8.00 (Borate), NIST Standard Solution ready to use for pH measurement

Specifications

Specifications

Colorless
Liquid
217672
8
2.5 L
HDPE plastic bottle
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Reproductive toxicity Category 1B

Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS Documents

Documents

Certificates
Special Offers

Special Offers