missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH7.00 (Phosphate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™

Brand:  Fisher Chemical J/2850/08

15.75 GBP valid until 2021-10-30
Use promo code "14806" to get your promotional price.



Code : 23

Additional Details : CAS Number : 7558-79-4 Weight : 0.50000kg

Packaging HDPE plastic bottle
Quantity 500mL

 View more versions of this product

Product Code. 10082521

  • £28.24 / 500mL

In stock, ready to ship: 256

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Buffer Solution pH7.00 (Phosphate), NIST Standard Solution ready to use for pH measurement

Ready to use for pH measurement.
Each solution is easily identified by its color, saving time and reducing errors.
Our buffer solutions are a high quality and reliable range of products that ensure accurate results for calibration/titration.
  • Standard and color-coded buffer solutions
  • Manufactured to a tolerance of ±0.02 pH units
  • Accurately standardized and labelled with lot-specific pH at 20°C
  • All buffers are traceable to NIST and BS 1647 standard
  • Packed in HDPE containers to help protect the products integrity an ensure safe handling (refers to 1L, 2.5L, and 5L quantities)
  • Standard buffers are available over the range pH2 to pH10
  • Color-coded buffer solutions are available over the range pH4, pH7, and pH
Specifications

Chemical Identifiers

217672

Specifications

217672
HDPE plastic bottle
Colorless
Liquid
7
500mL
Videos
SDS Documents
Certificates