Learn More
Buffer solution pH 9.2, Borate buffer, traceable to NIST, ready to use, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 383875000
Code : 29
Additional Details : Weight : 0.50000kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Buffer solution pH 9.2 | |
1330-43-4,7732-18-5,7647-14-5 | |
500 mL | |
1 |
Borate buffer, traceable to NIST, ready to use | |
1.0000g/mL | |
MFCD00163147 | |
Solubility in water: miscible. |
RUO – Research Use Only