missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 9.180, Fisher Chemical™
Shop All Fisher Chemical Products

Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
Specifications
Specifications
| pH | 9.180 |
| Physical Form | Liquid |
| Quantity | 1000 mL |
| Solution Type | Buffer |
| Formulation | Tetraborate Sodium |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction