missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 9.180 (25°C), With fungicide, according to DIN 19266, Solstice

Product Code. 15624020
Change view
Click to view available options
Quantity:
6 x 1 L
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15624020 6 x 1 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15624020 Supplier Honeywell-Fluka Supplier No. 335956X1L
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

With fungicide, according to DIN 19266

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer solution pH 9.180 (25°C)
Name Note With fungicide, according to DIN 19266
CAS 1330-43-4, 7732-18-5
Boiling Point 100°C
CAS Min % >0.1000%, >99.0000%
CAS Max % <0.5000%, <100.0000%
Density 1.0 g/cm3
For Use With (Application) pH Calibration
For Use With (Equipment) pH Meter and Electrode
pH 9.1
Physical Form Liquid
Quantity 6 x 1 L
Solution Type Buffer
Packaging Poly Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.