missing translation for 'onlineSavingsMsg'
Learn More

Color Coded pH Buffers in Twin Neck Bottle Format, Reagecon™

Product Code. 11454599 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
pH:
10 at 20°C
10 at 25°C
7 at 20°C
7 at 25°C
Unit Size:
1 L
250mL
500mL
Product Code. Quantity pH unitSize
11454599 1 L 10 at 25°C 1 L
11434599 250 mL 10 at 25°C 250mL
11454589 250 mL 10 at 20°C 250mL
11404599 250 mL 7 at 25°C 250mL
12037365 250 mL 7 at 20°C 250mL
11444599 500 mL 10 at 25°C 500mL
11802172 500 mL 7 at 25°C 500mL
11464589 1 L 10 at 20°C 1 L
11444589 1 L 7 at 20°C 1 L
11424599 1 L 7 at 25°C 1 L
10 options
This item is not returnable. View return policy

Product Code. 11454599

Brand: Reagecon 110025CTT

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use color coded pH buffers for ease of use. Reagecon™ Color Coded pH Buffer in a Twin Neck Bottle Format are tested at 20°C or 25°C and are certified and NIST traceable.

  • Colored pH Buffer Solutions
  • Tested at 20°C or 25°C 
  • Certified by Reagecon’s ISO/IEC 17025 AccreditedTest Method
  • NIST traceable
  • Presented in various pack sizes
Twin Neck Bottle Advantages
  • No possibility of contamination
  • No need for separate measuring container for use in the calibration of the Electrode
  • Correct quantity of buffer required for calibration is dispensed into the calibrating chamber giving rise to no waste
  • Ideally suited for field work
  • Easy to carry
  • 250ml, 500ml and 1L sizes available

Specifications

Color Blue
pH 10 at 25°C
Quantity 1 L
Packaging HDPE Twin Neck Bottle
compliance-icons
Product Identifier
  • BUFFER SOLUTION pH 10.00 ± 0.01 @ 25°C
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Boric Acid, DisodiumSalt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.