Learn More
Buffer solution pH 7.00 (+/-0.024 @ 25oC) Colored Green Specpure NIST Traceable
Description
Buffer solutions play a substantial role in industrial and physiological processes. Buffer solutions of known and precise pH value allow to set the testing system to measure the samples of unknown pH more precisely and accurately.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Buffer solution |
| Name Note | NIST Traceable |
| CAS | 1336-21-6 |
| Color | Green |
| Physical Form | Liquid |
| Quantity | 1 L |
| Molecular Formula | H5NO |
| MDL Number | MFCD00066650 |
| Traceability to NIST | Yes |
| Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
| Vis mere |
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.