missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 7, Mettler Toledo™

Product Code. 12808311 Shop All Mettler Toledo Products
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12808311 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12808311 Supplier Mettler Toledo Supplier No. 51340229

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Compatible with EasyPlus™ Titration Sensors.

  • Provided with a quality test certificates that guarantees the specifications and compliance with international standards
TRUSTED_SUSTAINABILITY

Specifications

Color Green
pH 7
Physical Form Liquid
Quantity 1000 mL
Solution Type pH Buffer
Odor None
Packaging 1000mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.