Learn More
Buffer solution pH 7.00 (+/-0.024 @ 25oC) No Color Specpure NIST Traceable, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 038712.K2
33.35 GBP valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Code : NEW
Additional Details : CAS Number : 1336-21-6 Weight : 0.01000kg
Description
Used to determine pH of solutions in various industrial applications. Buffer solutions of known and accurate pH value allows to adjust testing system to measure the samples with unknown pH more precisely and accurately.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Buffer solution | |
Colorless | |
Liquid | |
H5NO | |
MFCD00066650 | |
pH 7.00 buffers are in solution form. Miscible with water. | |
N.O | |
35.05 | |
CHEBI:18219 |
1336-21-6 | |
1.004-1.005g/cm3 at 20°C | |
1 L | |
Odorless | |
Yes | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
azanium;hydroxide | |
14923 | |
Specpure™ |
RUO – Research Use Only