missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 7.0 (20°C), Potassium Dihydrogen Phosphate / Disodium Hydrogen Phosphate, Traceable to SRM from NIST, with Fungicide, Honeywell Fluka™

Product Code. 15644030 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
5 L
6 x 100 mL
6 x 500 mL
10 L
6 x 1 L
Unit Size:
1 L
10 L
100mL
5 L
500mL
Pack of 6
This item is not returnable. View return policy

Product Code. 15644030

Brand: Honeywell-Fluka 3364610LVP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potassium dihydrogen phosphate / disodium hydrogen phosphate, traceable to SRM from NIST, with fungicide.

Leading the Industry For Over 65 Years. Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured.

Composition per liter:

  • 3.52 g Potassium dihydrogen phosphate (KH2PO4)
  • 7.26 g di-Sodium hydrogen phosphate-2-hydrate (Na2HPO4*2H20)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
PubChem CID 14923
ChEBI CHEBI:18219
SMILES N.O

Specifications

pH 7.0
Quantity 10 L
MDL Number MFCD00066650
UN Number NONH for all modes of transport
IUPAC Name azanium;hydroxide
Chemical Name or Material Buffer solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.