missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 6.0 (20°C), Citric acid / sodium hydroxide solution, with fungicide, traceable to SRM from NIST, Honeywell Fluka™

Product Code. p-7122059 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
6 x 1 L
Unit Size:
1 L
Pack of 6
This item is not returnable. View return policy

Product Code. 15664000

Brand: Honeywell-Fluka 335451L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Citric acid / sodium hydroxide solution, with fungicide, traceable to SRM from NIST

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer solution pH 6.0 (20°C)
Name Note Citric acid / sodium hydroxide solution, with fungicide, traceable to SRM from NIST
Boiling Point 100°C
CAS Min % ≥98.0000%
CAS Max % ≤99.0000%, ≤2.0000%
CAS 68-04-2, 144-33-2
Density 1.00 g/cm3
Quantity 1 L
Solution Type Buffer
MDL Number MFCD00012462
Packaging Poly Bottle
Synonym Sodium citrate buffer solution, Buffer solution from sodium citrate
Show More Show Less
Product Identifier
  • Buffer solution pH 6.0 (20 °C)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.