missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 6.0 (20°C), Citric acid / sodium hydroxide solution, with fungicide, traceable to SRM from NIST, Honeywell Fluka™

Product Code. 15674000 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
Unit Size:
1 L
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15674000 6 x 1 L Pack of 6
15664000 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15674000 Supplier Honeywell-Fluka Supplier No. 335456X1L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Citric acid / sodium hydroxide solution, with fungicide, traceable to SRM from NIST

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer solution
Name Note Citric acid/sodium hydroxide solution, with fungicide, traceable to SRM from NIST
CAS 1336-21-6
Quantity 6 x 1 L
Molecular Formula H5NO
MDL Number MFCD00066650
Packaging Poly bottle
UN Number NONH for all modes of transport
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILES N.O
IUPAC Name azanium;hydroxide
Molecular Weight (g/mol) 35.05
PubChem CID 14923
ChEBI CHEBI:18219
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.