missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 5.5, Fisher Chemical™
Click to view available options
Quantity:
1000 mL
Specifications
pH | 5.5 |
Physical Form | Liquid |
Quantity | 1000 mL |
Solution Type | Buffer |
Formulation | Phosphate - Citrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'