missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution, pH 5.2 EU Pharmacopoeia, Fisher Chemical™
Shop All Fisher Chemical Products

Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
Specifications
Specifications
| CAS | 1336-21-6 |
| pH | 5.2 |
| Quantity | 1000 mL |
| Molecular Formula | H5NO |
| MDL Number | MFCD00066650 |
| Packaging | Bottle |
| InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
| SMILES | N.O |
| IUPAC Name | azanium;hydroxide |
| Molecular Weight (g/mol) | 35.05 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction