missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 4, Red, Reagecon™
BIOPHARMA

Product Code. 11474589 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
5 L
Packaging:
Bag-in-Box Container
HDPE Bottle GL45
HDPE Twin Neck Bottle
pH:
4 at 20°C
4 at 25°C
Unit Size:
1 L
250mL
5 L
500mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging pH unitSize
11474589 250 mL HDPE Twin Neck Bottle 4 at 25°C 250mL
11414589 250 mL HDPE Twin Neck Bottle 4 at 20°C 250mL
11860513 500 mL HDPE Bottle GL45 4 at 25°C 500mL
11858783 500 mL HDPE Twin Neck Bottle 4 at 25°C 500mL
11424589 1 L HDPE Twin Neck Bottle 4 at 20°C 1 L
11494589 1 L HDPE Twin Neck Bottle 4 at 25°C 1 L
11494619 5 L Bag-in-Box Container 4 at 20°C 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 11474589 Supplier Reagecon Supplier No. 1040225CTT

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Colored pH Buffer solutions in Twin-neck containers. Reagecon™ Buffer Solution pH 4, Red are ISO 17025 accredited and NIST traceable.

  • Colored
  • Twin-neck containers with integrated calibrating chamber
  • Tested at 20°C or 25°C
  • Certified by ISO 17025 Accredited Test Method
  • NIST traceable
  • Presented in various pack sizes

Specifications

Color Red
pH 4 at 25°C
Quantity 250 mL
Packaging HDPE Twin Neck Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.