missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 4, Citrate buffer

Product Code. 10388803 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
5 L
Unit Size:
1 L
5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10388803 5 L 5 L
10661832 500 mL 500mL
10349240 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10388803 Supplier Thermo Scientific Acros Supplier No. 258580050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68-04-2 | C8H5KO4

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

pH buffer
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer solution pH 4
Name Note Phthalate buffer
CAS 68-04-2, 7647-14-5, 7732-18-5
Color Colorless
pH 3.98 to 4.02 (20°C)
Physical Form Liquid
Quantity 5 L
Molecular Formula C8H5KO4
MDL Number MFCD00130806
Packaging Plastic bottle
Traceability to NIST Traceable to NIST
Solubility Information Solubility in water: soluble.
Formula Weight 204.23
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.