missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Reagecon's pH Buffer Solution at 20°C is tested by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable, has guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
Specifications
| pH | 4.10 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Packaging | HDPE Bottle GL45 |
| Chemical Name or Material | Low Ionic Strength Buffer Solution |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?