missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 2, Mettler Toledo™
Brand: Mettler Toledo 51319011
| Quantity | 1000mL |
|---|---|
| Packaging | 1000mL |
This item is not returnable.
View return policy
Description
Specifications
| 4.64 | |
| pH Buffer Solution | |
| 1000mL |
| 1000mL | |
| None |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Buffer pH 2.00
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contain: Citric acid anhydrous, Hydrogen chlorideanhydrous
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?