missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 2.00 (Glycine), NIST Standard Solution Ready to Use for pH Measurement, Fisher Chemical™

Product Code. 10588040 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Unit Size:
1 L
2.5 L
500mL
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Product Code. 10588040

Brand: Fisher Chemical J/2800/17

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Buffer Solution pH 2.00 (Glycine), NIST Standard Solution Ready to Use for pH Measurement

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number 217672

Specifications

Color Colorless
pH 2
Physical Form Liquid
Quantity 2.5 L
Packaging HDPE plastic bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.