missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution Ph 2.0 (20 °C), Honeywell Fluka™

Product Code. 15633180 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mL
1 L
6 x 1 L
Unit Size:
1 L
100mL
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15633180 100 mL 100mL
15643180 1 L 1 L
15653180 6 x 1 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15633180 Supplier Honeywell-Fluka Supplier No. 31045100ML
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Citric acid / hydrochloric acid / sodium chloride, with fungicide, traceable to SRM from NIST and PTB

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer Solution Ph 2.0 (20 °C)
Name Note With fungicide
CAS 7732-18-5, 7647-14-5, 77-92-9
Quantity 100 mL
MDL Number MFCD00146206
Packaging Plastic Bottle
UN Number 1789
compliance-icons
Product Identifier
  • Buffer solution pH 2.0 (20 °C)
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: Citric acid, Sodiumchloride, hydrochloric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.