missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution, pH 13.00 (+/-0.026 @ 25°C), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 046307.K2

Code : NEW

Additional Details : Weight : 1.00000kg


 View more versions of this product

Product Code. 15464035

  • £28.90 / 1 L
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Buffer solution, pH 13.00 (±0.01 at 25°C), No Color, Specpure™, NIST Traceable is used as a means of keeping the pH at a nearly constant value in different chemical applications. Buffer solution is used in alcoholic fermentation, leather tanning, electroplating, organic synthesis, sugar and paper manufacturing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Buffer solution, pH 13.00 (±0.01 at 25°C)
1 L
MFCD03458582
Miscible with water
NIST Traceable
Colorless
Specpure™
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Buffer solution, pH 13.00

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Sodium hydroxide

SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only