missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 12.00, Reagecon™
BIOPHARMA

Product Code. 11434569 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434569 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11434569 Supplier Reagecon Supplier No. 112025
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Certified by an ISO 17025 accredited test method and fully NIST traceable. Reagecon™ Buffer Solution, pH 12.00 is tested at 20°C or 25°C to an accuracy level of ±0.05. This calibration and control buffer is specially formulated to ensure its stability, even after opening the bottle.

To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.

  • Traceable to SRM for NIST
  • Certification to ISO 9001:2008
  • Accredited to ISO 17025

Specifications

pH 12
Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.