missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 11.00, Reagecon™

Certified by an ISO 17025 accredited test method and fully NIST traceable. Reagecon™ Buffer Solution, pH 11.00 is tested at 20°C to an accuracy level of ±0.05. This calibration and control buffer is specially formulated to ensure its stability, even after opening the bottle.

Brand:  Reagecon 11105

Code : 23

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11464529

  • £33.35 / 500mL
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.

  • Traceable to SRM for NIST
  • Certification to ISO 9001:2008
  • Accredited to ISO 17025
Specifications

Specifications

0°C
Colorless
Liquid
pH Buffer
Yes
100°C
11.00 ±0.05
500 mL
Characteristic
Fully miscible in water
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers