missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution, pH 11.00 (+/-0.026 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 042424.AP

Code : NEW

Additional Details : CAS Number : 1336-21-6 Weight : 0.01000kg

 View more versions of this product

Product Code. 11306099

  • £18.40 / 500mL
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Buffer solution (pH 11) is used as a means of keeping pH at a nearly constant value in different chemical applications. Buffer solution is used in alcoholic fermentation, leather tanning, electroplating, organic synthesis, sugar and paper manufacturing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Buffer solution
Colorless
500 mL
Odorless
Miscible with water.
N.O
35.05
CHEBI:18219
1336-21-6
Liquid
H5NO
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
amine hydrate
14923
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only