missing translation for 'onlineSavingsMsg'
Learn More

Color Coded pH Buffer Solution Tested at 20°C, Reagecon™
BIOPHARMA

Product Code. 11486424 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Color:
Blue
Red
Yellow
Unit Size:
1 L
500mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Color unitSize
11486424 500 mL Blue 500mL
12808861 500 mL Red 500mL
12201473 500 mL Yellow 500mL
11404579 1 L Blue 1 L
11464569 1 L Red 1 L
11484569 1 L Yellow 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11486424 Supplier Reagecon Supplier No. 11005C

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Use color coded pH buffers for ease of use. Reagecon™ Color Coded pH Buffer Solutions Tested at 20°C are certified and NIST traceable.

  • Colored pH Buffer Solutions
  • Tested at 20°C 
  • Certified by Reagecon’s ISO/IEC 17025 AccreditedTest Method
  • NIST traceable
  • Presented in various pack sizes

Specifications

Color Blue
pH 10
Physical Form Liquid
Quantity 500 mL
compliance-icons
Product Identifier
  • BUFFER SOLUTION pH 10.00 ± 0.01 @ 20°C
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: Boric Acid, DisodiumSalt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.