missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 10, Mettler Toledo™
35.22 GBP valid until 2024-12-31
Use promo code "23284" to get your promotional price.
Use promo code "23284" to get your promotional price.
Click to view available options
Quantity:
250 mL
Packaging:
1 Pk.
6 Pk.
Specifications
Color | Colorless |
pH | 10 |
Physical Form | Liquid |
Quantity | 250 mL |
Solution Type | pH Buffer |
Formulation | 95 to 99% Deionized water, 0.1 to 0.5% Sodium hydroxide; caustic soda |
Odor | None |
Packaging | 1 Pk. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'