missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 10.00 (20°C), Fluka™

Product Code. 16232892 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 16232892

Brand: Honeywell-Fluka 35083500ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Certified, colored blue

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer Solution pH 10.00 (20°C)
CAS 7732-18-5, 1310-73-2, 12179-04-3
Melting Point <0°C
Boiling Point 100°C (1.013 hPa)
Color Blue
pH 10
Quantity 500 mL
Packaging Plastic Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.