missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 1.68 at 25 °C, Reagecon
Guaranteed stability throughout entire shelf life even after opening the bottle, NIST traceable
Brand: Reagecon 1016825
Additional Details : Weight : 1.00000kg
Description
- Tested by ISO 17025 accredited test method
Specifications
Buffer solution | |
Liquid | |
HDPE bottle GL45 |
1.68 ±0.01 | |
1 L | |
Yes |