Learn More
Buffer solution, pH 1.68 (+/-0.026 @ 25°C), No Color, Specpure, NIST Traceable
Description
Buffer Solution pH 1.68 used as a means of keeping pH at a nearly constant value in a different chemical applications. Glycine buffer solution pH 2 used for eluting antibodies in various antibody purification procedures i.e. protein A-agarose, protein G-agarose, affinity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Buffer solution |
| CAS | 7447-40-7, 7647-01-0, 7732-18-5 |
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 4 L |
| Odor | Odorless |
| MDL Number | MFCD00147253 |
| Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
| Solubility Information | Miscible with water. |
| Concentration or Composition (by Analyte or Components) | Potassium chloride: 0.7%; Hydrochloric acid: 0.7%; water: 98.6% |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.