missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Contains potassiumchloride / hydrochloric acid
- This buffer is traceable to NIST Standard Reference Buffer
Specifications
Specifications
| pH | 1.00 ±0.02 |
| Quantity | 1 L |
Safety and Handling
Product Identifier
- Buffer Solution, pH 1.00
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: hydrochloric acid
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?