missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution, KCl for AAS 0.02mg/L HCl matrix, Fisher chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Specifications
Specifications
| Concentration | 0.02 mg/L |
| Solvent or Matrix | HCl |
| Quantity | 100 mL |
| Chemical Name or Material | Buffer solution, KCl for AAS |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction