missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution for HPCE, 50 mM sodium phosphate, pH 7.0 (25 °C), Honeywell Fluka™

Product Code. 15697250 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Plastic bottle
Unit Size:
100mL
50mL
This item is not returnable. View return policy

Product Code. 15697250

Brand: Honeywell-Fluka 8263650ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Absorbance 1: 0 to 0.02; 2: 0 to 0.05; 3: 0 to 0.25
Chemical Name or Material Buffer solution HPCE pH 7.0
Color Colorless
Concentration 50 mM
pH 7.0
Physical Form Liquid
Quantity 50 mL
Packaging Plastic bottle
UN Number NONH for all modes of transport
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.