missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution for HPCE, 50 mM sodium phosphate, pH 7.0 (25 °C), Honeywell Fluka™

Product Code. 15687250 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Plastic Bottle
Unit Size:
100mL
50mL
This item is not returnable. View return policy

Product Code. 15687250

Brand: Honeywell-Fluka 82636100ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Absorbance max. 0.02 at 250 nm, max. 0.05 at 230 nm, max. 0.3 at 210 nm
Chemical Name or Material Buffer solution for HPCE
Name Note 50 mM sodium phosphate, pH 7.0 (25 °C)
CAS Min % ≥93.7600%, ≥2.5200%
CAS Max % ≤94.3400%, ≤3.1000%
Color Liquid
CAS 1310-73-2, 7664-38-2
Concentration 50 mM
pH 7.0
Quantity 100 mL
Assay Percent Range 6.9 to 7.1%
Linear Formula 2-(C6H5NH)C6H4CO2H
MDL Number MFCD00002421
Packaging Plastic Bottle
UN Number NONH for all modes of transport
Synonym Sodium phosphate buffer solution, Buffer solution from phosphoric acid/di-Sodium hydrogen phosphate, Phosphoric acid/di-Sodium hydrogen phosphate solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.