missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution, Borax/Hydroxide Sodium, pH 10.00, Fisher Chemical™
Shop All Fisher Chemical Products

Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Specifications
Specifications
| Chemical Name or Material | Buffer Solution, Coloured Borax/Sodium Hydroxide |
| pH | 10 |
| Quantity | 500 mL |
| Packaging | Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction