missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution, pH 7.00 (+/-0.024 @ 25oC), Colored Yellow, Specpure, NIST Traceable

Product Code. 11338398
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Unit Size:
1 L
4 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338398 4 L 4 L
11450271 500 mL 500mL
11348398 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11338398 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' 040439.K7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1310-73-2

Used to determine pH of solutions in various industrial applications. Buffer solutions of known and accurate pH value allows to adjust testing system to measure the samples with unknown pH more precisely and accurately.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer solution
CAS 1310-73-2, 7778-77-0, 7732-18-5
Color Yellow
Physical Form Liquid
Quantity 4 L
MDL Number MFCD00146252
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Solubility Information Miscible with water.
Concentration or Composition (by Analyte or Components) Sodium hydroxide: 0.7%; Potassium dihydrogen phosphate: 0.7%; water: 98.6%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.