Få mere at vide
Buffer solution, pH 7.00 (+/-0.024 @ 25oC), Colored Yellow, Specpure, NIST Traceable
Beskrivelse
Used to determine pH of solutions in various industrial applications. Buffer solutions of known and accurate pH value allows to adjust testing system to measure the samples with unknown pH more precisely and accurately.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tekniske data
Tekniske data
| Chemical Name or Material | Buffer solution |
| CAS | 1310-73-2, 7778-77-0, 7732-18-5 |
| Color | Yellow |
| Physical Form | Liquid |
| Quantity | 500 mL |
| MDL Number | MFCD00146252 |
| Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
| Solubility Information | Miscible with water. |
| Concentration or Composition (by Analyte or Components) | Sodium hydroxide: 0.7%; Potassium dihydrogen phosphate: 0.7%; water: 98.6% |
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.