missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 4.00 (+/-0.022 @ 25oC) Colored Red Specpure NIST Traceable

Product Code. 11400171
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Unit Size:
1 L
20 L
4 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400171 20 L 20 L
11490161 500 mL 500mL
11378987 1 L 1 L
11388987 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11400171 Supplier Thermo Scientific Alfa Aesar Supplier No. 038713.M4
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1310-73-2

Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications. Buffer solutions are essential to maintain the suitable condition to keep the correct pH for enzymes in many organisms to work. It is also used in fermentation processes and in setting the correct conditions for dyes used in coloring fabrics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer solution
CAS 1310-73-2, 7778-77-0, 7732-18-5
Color Red
Physical Form Liquid
Quantity 20 L
MDL Number MFCD00146206
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Solubility Information Miscible with water.
Concentration or Composition (by Analyte or Components) Sodium hydroxide: 0.7%; Potassium dihydrogen phosphate: 0.7%; water: 98.6%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.