missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 13.0 (20°C), Glycine / sodium hydroxide / sodium chloride solution, Honeywell Fluka™

Product Code. 15604010 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 15604010

Brand: Honeywell-Fluka 335521L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Glycine / sodium hydroxide / sodium chloride solution

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 1336-21-6
pH 13.0
Quantity 1 L
Molecular Formula H5NO
MDL Number MFCD00066650
UN Number NONH for all modes of transport
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILES N.O
IUPAC Name azanium;hydroxide
Molecular Weight (g/mol) 35.05
PubChem CID 14923
ChEBI CHEBI:18219
Chemical Name or Material Buffer solution
Show More Show Less
compliance-icons
Product Identifier
  • Buffer solution pH 13.0 (20 °C)
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide; caustic soda
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.