missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution, 11.0, boric acid / sodium hydroxide solution / potassium chloride, Honeywell Fluka™

Product Code. 15654050 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
6 x 100 mL
6 x 500 mL
6 x 1 L
Unit Size:
1 L
100mL
500mL
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15654050 500 mL 500mL
15634050 100 mL 100mL
15664050 6 x 100 mL Pack of 6
15644050 1 L 1 L
15684050 6 x 500 mL Pack of 6
15674050 6 x 1 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15654050 Supplier Honeywell-Fluka Supplier No. 33650500ML
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Boric acid / sodium hydroxide solution / potassium chloride

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name azanium;hydroxide
SMILES N.O

Specifications

pH 11.0
Quantity 500 mL
UN Number NONH for all modes of transport
Chemical Name or Material Buffer solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.