Learn More
Fisherbrand™ Buffer Saver Blocks
Physically reduce volume in positive and negative compartments of gel chamber thus reducing buffer requirements.
Brand:  Fisherbrand™
Code : 46
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11867633
  • £32.55 / Pack of 2
Estimated Shipment: 10-07-2024
to see stock.
Add to basket
Description
Description
  • Economic low gel and buffer volumes
  • Available for multiSUB Horizontal Gel Units
Specifications
Specifications
Sub-Gel Midi Horizontal Gel Electrophoresis Unit
Buffer Saver Block
2 Each