missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Buffer S1

Product Code. 10556721
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10556721 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10556721 Supplier Macherey-Nagel™ Supplier No. 740516.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Replacement component for use in NucleoBond™ Plasmid purification kits. Macherey-Nagel™ Buffer S1 is a resuspension buffer that does not contain RNase A.

NucleoBond™ Plasmid purification kits incorporate NucleoBond™ AX technology, a silica-based anion exchanger developed for routine separation of different classes of nucleic acids, allowing superior biological activity of the purified nucleic acids and rendering high mass recoveries possible by minimising non-specific adsorption effects.

Kit types include: NucleoBond™ Mini (PC 20), NucleoBond™ Midi (PC 100), NucleoBond™ Maxi (PC 500), NucleoBond™ Mega (PC 2000), and NucleoBond™ Giga (PC 10,000).

  • NucleoBond™ folded filters eliminate the centrifugation step for easy, fast and convenient clarification of bacterial lysate
  • Columns run by gravity flow, allowing purification of multiple samples in parallel
  • Working range from nanograms to milligrams DNA
  • Recovery of plasmid DNA is >90%
  • Transfection-grade DNA
  • High batch-to-batch reproducibility
  • No use of organic solvents
  • Alcohol precipitation of DNA/RNA High and low copy number plasmid DNA purifaction from E. coli that is suitable for transfection, sequencing and cloning.

Specifications

Chemical Name or Material Buffer S1
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.