missing translation for 'onlineSavingsMsg'
Learn More

Buffer R1 solution, pH 9 EU Pharmacopoeia, Fisher Chemical™
BIOPHARMA

Product Code. 12910744 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
This item is not returnable. View return policy

Product Code. 12910744

Brand: Fisher Chemical SOL 097 1L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Chemical Name or Material Buffer solution R1
pH 9
Quantity 1000 mL
Packaging Bottle
Grade Eu. Ph. Reference
compliance-icons
Product Identifier
  • Buffer solution R1 - pH 9.0 European Pharmacopoeia (Ph Eur) Ref: 4007100
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360D-May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : boric acid, sodium hydroxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.