missing translation for 'onlineSavingsMsg'
Learn More

Buffer Powder Pillows, Hach Pacific Pty Ltd

Product Code. 15618115
Change view
Click to view available options
Quantity:
250 Pc.
Color:
Blue
Red
Yellow
pH:
10.01 ±0.02
4.01 ±0.02
7.00 ±0.02
Unit Size:
Pack of 250
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Color pH Unit Size
15618115 250 Pc. Red 4.01 ±0.02 Pack of 250
15669524 250 Pc. Blue 10.01 ±0.02 Pack of 250
15628115 250 Pc. Yellow 7.00 ±0.02 Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15618115 Supplier Hach Supplier No. 2226964
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

pH Buffer standard solutions for laboratory and field analyses.

pH Buffer Powder Pillows for pH electrode calibration. Each powder pillow prepares 50 mL of fresh buffer solution. Colour-coded for easy identification.

  • Traceable to NIST standard reference materials
  • Design for use with Hach meters portfolio and other meters.
  • Reliable packaging

  • pH Buffer standard solutions for laboratory and field analyses
  • pH Buffer Powder Pillows for pH electrode calibration. Each powder pillow prepares 50 mL of fresh buffer solution. Colour-coded for easyidentification.
  • Traceable to NIST standard reference materials
  • Whether in the laboratory or in the field, always perform measurement results you can trust is essential. The quality of the standard pH that are used are key.
  • Design for use with Hach meters portfolio and other meters.
  • The basis for accurate pH measurements is good care and calibration of the pH Electrode.
  • Reliable packaging
  • Supplied in an airtight packaging ensuring guaranteed shelf-life.

    Specifications

  • Color coded?: Yes
  • Description: pH Buffer Calibration reagent
  • NIST Traceable: Yes
  • Package Type: Powder Pillows
  • Shelf Life: 24 months
  • Special Remarks: Discard
  • prepared solution after use
  • Storage conditions: 10 - 25 °C (protect from light)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium hydrogen phthalate
CAS 877-24-7
Melting Point 295°C to 300°C (decomposition)(literature)
Boiling Point 98.5°C to 99.5°C (740 mmHg) (literature)
Color Red
pH 4.01 ±0.02
Physical Form Powder
Quantity 250 Pc.
Molecular Formula C8H5KO4
Packaging Powder Pillows
Molecular Weight (g/mol) 204.22
Show More Show Less
compliance-icons
Product Identifier
  • Buffer Powder Pillows pH 4.01
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362-Take off contaminated clothing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: potassium hydrogen phthalate, SPADNS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.