missing translation for 'onlineSavingsMsg'
Learn More

Buffer, pH 10, Blue, Reagecon™
BIOPHARMA

Product Code. 11414609 Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 11414609

Brand: Reagecon H100525C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Suitable for use where very accurate measurements are required. Reagecon Buffer, pH 10, Blue is a colored, high resolution, professional range buffer manufactured to exacting specifications with an extended shelf life.

Used with pH meters as calibration standards and as cover values that can be used as control standards for all pH applications.

  • Tested at 25°C and certified to ISO 17025 by an accredited test method
  • Supplied with a detailed Certificate of Analysis which outlines traceability to N.I.S.T
  • Temperature dependence data is printed on the label, as are lot numbers and expiry dates
  • Color coded: Blue (pH 10)

Specifications

Melting Point 0°C
Boiling Point 100°C
Color Blue
Certificate of Analysis Supplied
Density 1.02913g/cm3
pH 10.000 ±0.010
Physical Form Liquid
Quantity 500 mL
Formulation ≤2.5% Boric acid, disodium salt
Packaging Bottle
Traceability to NIST Traceable to NIST
Vapor Pressure 23hPa (at 20°C)
Solubility Information Fully miscible
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.